4-(2-Fluorophenyl)-2-methylthiazole 96%
Supplier: Apollo Scientific
PC910849-10G
PC910849-1G
PC910849-5G
APOSPC910849-10GEA
502
EUR
APOSPC910849-10G
APOSPC910849-1G
APOSPC910849-5G
4-(2-Fluorophenyl)-2-methylthiazole 96%
4-(2-Fluorophenyl)-2-methylthiazole
Formula:
C₁₀H₈FNS Storage Temperature: Ambient |
MDL Number:
MFCD21333097 CAS Number: 1355248-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...