2-[4-(4-Fluorobenzoyl)phenyl]-2-(4-fluorophenyl)acetonitrile 97%
Supplier: Apollo Scientific
PC9320-1G PC9320-500MG PC9320-5G
APOSPC9320-1GEA 203 EUR
APOSPC9320-1G APOSPC9320-500MG APOSPC9320-5G
2-[4-(4-Fluorobenzoyl)phenyl]-2-(4-fluorophenyl)acetonitrile 97%
2-[4-(4-Fluorobenzoyl)phenyl]-2-(4-fluorophenyl)acetonitrile
Formula: C₂₁H₁₃F₂NO
MW: 333.331 g/mol
Melting Pt: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00232016
CAS Number: 339115-12-7