2-[4-(4-Fluorobenzoyl)phenyl]-2-phenylacetonitrile 97%
Supplier: Apollo Scientific
PC9329-1G PC9329-500MG PC9329-5G
APOSPC9329-1GEA 302 EUR
APOSPC9329-1G APOSPC9329-500MG APOSPC9329-5G
2-[4-(4-Fluorobenzoyl)phenyl]-2-phenylacetonitrile 97%
2-[4-(4-Fluorobenzoyl)phenyl]-2-phenylacetonitrile
Formula: C₂₁H₁₄FNO
MW: 315.34 g/mol
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00231980
CAS Number: 338791-81-4