3-Bromo-5-(trifluoromethoxy)toluene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-5-methyl-α,α,α-trifluoroanisole, 3-Bromo-5-methylphenyl trifluoromethyl ether, 1-Bromo-3-methyl-5-(trifluoromethoxy)benzene
PC9926-1G PC9926-5G
APOSPC9926-1GEA 72.5 EUR
APOSPC9926-1G APOSPC9926-5G
3-Bromo-5-(trifluoromethoxy)toluene 97%
3-Bromo-5-(trifluoromethoxy)toluene
Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368791
CAS Number: 887266-91-3