Kinetin
Supplier: Apollo Scientific
Warning
BI5193-25G BI5193-5G BI5193-100G
APOSBI5193-25GEA 29 EUR
APOSBI5193-25G APOSBI5193-5G APOSBI5193-100G
Kinetin
Kinetin
  • White to off-white powder
  • Plant tissue culture tested
Formula: C₁₀H₉N₅O
Melting Pt: 264…270 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00075757
CAS Number: 525-79-1
EINECS: 208-382-2