Filipin III (from Streptomyces filipinesis)
Supplier: Apollo Scientific
Warning
BIA4324-1MG BIA4324-5MG
APOSBIA4324-1MGEA 157 EUR
APOSBIA4324-1MG APOSBIA4324-5MG
Filipin III (from Streptomyces filipinesis)
Filipin III
Storage Temperature: Freezer
MDL Number: MFCD00466923
CAS Number: 480-49-9