L-Leucine(4-methyl-7-coumarinylamide) hydrochloride
Supplier: Apollo Scientific
Warning
BIB1441-100MG BIB1441-250MG
APOSBIB1441-100MGEA 72.5 EUR
APOSBIB1441-100MG APOSBIB1441-250MG
L-Leucine(4-methyl-7-coumarinylamide) hydrochloride
L-Leucine(4-methyl-7-coumarinylamide) hydrochloride
Formula: C₁₆H₂₀N₂O₃·HCl
Melting Pt: 283 °C
Storage Temperature: Freezer
MDL Number: MFCD00043262
CAS Number: 62480-44-8