Adenosine 5'-diphosphate disodium salt (ADP disodium salt) ≥96%
Supplier: Apollo Scientific
Synonyms: Adenosine 5'-Diphosphate Disodium Salt, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate
BIB3001-10G BIB3001-5G
APOSBIB3001-10GEA 120 EUR
APOSBIB3001-10G APOSBIB3001-5G
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) ≥96%
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Formula: C₁₀H₁₃N₅Na₂O₁₀P₂
MW: 471.167 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00066635
CAS Number: 16178-48-6
EINECS: 240-314-7