L(+)-Lactic acid 98.0+%
Supplier: Apollo Scientific
Warning
Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid
BIB6089-100G BIB6089-50G BIB6089-25G
APOSBIB6089-100GEA 284 EUR
APOSBIB6089-100G APOSBIB6089-50G APOSBIB6089-25G
L(+)-Lactic acid 98.0+%
L(+)-Lactic acid
Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 235
Melting Pt: 53 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 201-196-2
Merck Index: 13,05352