Thaxtomin A
Supplier: Apollo Scientific
|
Warning
|
BIBR1102-1MG
BIBR1102-5MG
BIBR1102-500UG
APOSBIBR1102-1MGEA
284
EUR
APOSBIBR1102-1MG
APOSBIBR1102-5MG
APOSBIBR1102-500UG
Thaxtomin A
Thaxtomin A
Formula:
C₂₂H₂₂N₄O₆ Storage Temperature: Freezer |
CAS Number:
122380-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...