Cephalexin monohydrate
Supplier: Apollo Scientific
Danger
Synonyms: 7-(D-α-amino-phenylacetamido)-3-methyl-3-cepheme-4-carboxylic acid monohydrate, (6R,7R)-7-((R)-2-Amino-2-phenylacetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate
BIC0110-10G BIC0110-5G
APOSBIC0110-10GEA 71.2 EUR
APOSBIC0110-10G APOSBIC0110-5G
Cephalexin monohydrate
Cephalexin monohydrate
Formula: C₁₆H₁₇N₃O₄S
MW: 347.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00167148
CAS Number: 23325-78-2