Citreoviridin
Supplier: Apollo Scientific
|
Danger
|
BIC0127-1MG
BIC0127-5MG
APOSBIC0127-1MGEA
154
EUR
APOSBIC0127-1MG
APOSBIC0127-5MG
Citreoviridin
Citreoviridin
Formula:
C₂₃H₃₀O₆ Melting Pt: 110 °C Storage Temperature: Freezer |
CAS Number:
25425-12-1 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...