Cercosporin
Supplier: Apollo Scientific
BIC1090-1MG
BIC1090-5MG
APOSBIC1090-1MGEA
105
EUR
APOSBIC1090-1MG
APOSBIC1090-5MG
Cercosporin
Cercosporin
Formula:
C₂₉H₂₆O₁₀ Storage Temperature: Freezer |
CAS Number:
35082-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...