Concanamycin A

Supplier: Apollo Scientific

BIC1091-1MG BIC1091-100UG
APOSBIC1091-1MGEA 870 EUR
APOSBIC1091-1MG APOSBIC1091-100UG
Concanamycin A
Concanamycin A
Formula: C₄₆H₇₅NO₁₄
MW: 866.1 g/mol
Storage Temperature: Freezer
CAS Number: 80890-47-7
UN: 3462
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR