EDTA (ethylenediamine tetraacetic acid) ≥99%
Supplier: Apollo Scientific
Warning
Synonyms: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
BIE0729-1KG BIE0729-500G
APOSBIE0729-1KGEA 54.2 EUR
APOSBIE0729-1KG APOSBIE0729-500G
EDTA (ethylenediamine tetraacetic acid) ≥99%
EDTA (Ethylenediamine tetraacetic acid)
Formula: C₁₀H₁₆N₂O₈
MW: 292.25 g/mol
Boiling Pt: 614
Melting Pt: 245 °C (dec.)
Density: 1.46 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
Merck Index: 13,03546