Fast blue B salt zinc double salt
Supplier: Apollo Scientific
Danger
BIF2001-10G BIF2001-5G BIF2001-25G
APOSBIF2001-10GEA 168 EUR
APOSBIF2001-10G APOSBIF2001-5G APOSBIF2001-25G
Fast blue B salt zinc double salt
Fast Blue B salt zinc double salt
Formula: C₁₄H₁₂Cl₄N₄O₂Zn
Storage Temperature: Refrigerator
MDL Number: MFCD00075203
CAS Number: 14263-94-6
EINECS: 238-153-2