D-Tagatose
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(3S,4S,5R)-1,3,4,5,6-Pentahydroxyhexan-2-one
BIFS0115-100G
BIFS0115-250G
BIFS0115-50G
APOSBIFS0115-100GEA
183
EUR
APOSBIFS0115-100G
APOSBIFS0115-250G
APOSBIFS0115-50G
D-Tagatose
D-Tagatose
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00134449 CAS Number: 87-81-0 EINECS: 201-772-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...