HT-2 Toxin
Supplier: Apollo Scientific
|
Danger
|
BIH1001-1MG
BIH1001-5MG
APOSBIH1001-1MGEA
293
EUR
APOSBIH1001-1MG
APOSBIH1001-5MG
HT-2 Toxin
HT-2 Toxin
Formula:
C₂₂H₃₂O₈ Storage Temperature: Freezer |
CAS Number:
26934-87-2 UN: 3462 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...