K252a
Supplier: Apollo Scientific
Warning
BIK1001-1MG BIK1001-5MG
APOSBIK1001-1MGEA 109 EUR
APOSBIK1001-1MG APOSBIK1001-5MG
K252a
K252a
Melting Pt: 233…258 °C
Storage Temperature: Freezer
CAS Number: 99533-80-9