Ophiobolin A

Supplier: Apollo Scientific
Danger

BIM0137-1MG BIM0137-5MG
APOSBIM0137-1MGEA 241 EUR
APOSBIM0137-1MG APOSBIM0137-5MG
Ophiobolin A
Ophiobolin A
Formula: C₂₅H₃₆O₄
Storage Temperature: Freezer
MDL Number: MFCD00058400
CAS Number: 4611-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR