Dimidium bromide
Supplier: Apollo Scientific
Warning
Synonyms: 2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide
BIP4354-250MG BIP4354-1G BIP4354-5G
APOSBIP4354-250MGEA 36.4 EUR
APOSBIP4354-250MG APOSBIP4354-1G APOSBIP4354-5G
Dimidium bromide
Dimidium bromide
Formula: C₂₀H₁₈BrN₃
Melting Pt: 242…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00011757
CAS Number: 518-67-2
EINECS: 208-256-7