Mono(boc)(PEO)-3-diamine, boc-DOOA 98%
Supplier: Apollo Scientific
Warning
BIPG1810-1G BIPG1810-5G
APOSBIPG1810-1GEA 87 EUR
APOSBIPG1810-1G APOSBIPG1810-5G
Mono(boc)(PEO)-3-diamine, boc-DOOA 98%
Mono(boc)(PEO)-3-diamine, boc-DOOA
Formula: C₁₁H₂₄N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD03788155
CAS Number: 153086-78-3