Tetraethylene glycol bis(p-toluenesulphonate)
Supplier: Apollo Scientific
Synonyms: Bis[2-[2-(p-toluenesulphonyloxy)ethoxy]ethyl] Ether, ((Oxybis(ethane-2,1-diyl))bis(oxy))bis(ethane-2,1-diyl) bis(4-methylbenzenesulphonate), Bis-Tos-PEG4, Bis[2-(2-tosyloxyethoxy)ethyl] Ether, Tetraethylene Glycol Di-p-tosylate
BIPG1869-10G BIPG1869-5G BIPG1869-25G
APOSBIPG1869-10GEA 157 EUR
APOSBIPG1869-10G APOSBIPG1869-5G APOSBIPG1869-25G
Tetraethylene glycol bis(p-toluenesulphonate)
Tetraethylene glycol bis(p-toluenesulphonate)
Formula: C₂₂H₃₀O₉S₂
MW: 502.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075239
CAS Number: 37860-51-8