Gellan gum, powder
Supplier: Apollo Scientific
Warning
BIPG434-100G BIPG434-1KG BIPG434-500G
APOSBIPG434-100GEA 94.4 EUR
APOSBIPG434-100G APOSBIPG434-1KG APOSBIPG434-500G
Gellan gum, powder
Gellan gum
Melting Pt: >70 °C
Storage Temperature: Ambient
MDL Number: MFCD00131909
CAS Number: 71010-52-1