Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine)dihydrate
Supplier: Apollo Scientific
Danger
Synonyms: Spermine Dihydrate
BIS5944-25G BIS5944-5G
APOSBIS5944-25GEA 418 EUR
APOSBIS5944-25G APOSBIS5944-5G
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine)dihydrate
Spermine (N,N'-Bis(3-aminopropyl)-1,4-butanediamine) dihydrate
Formula: C₁₀H₂₆N₄·2H₂O
Melting Pt: 63…64 °C
Storage Temperature: Refrigerator
CAS Number: 403982-64-9
UN: 3259
ADR: 8 II