L(+)-Valine ≥99.5%
Supplier: Apollo Scientific
Warning
Synonyms: H-Val-OH, Valine, (S)-(+)-Valine
BIV0722-100G BIV0722-1KG
APOSBIV0722-100GEA 21.9 EUR
APOSBIV0722-100G APOSBIV0722-1KG
L(+)-Valine ≥99.5%
L(+)-Valine
USP, pyrogen free
Formula: C₅H₁₁NO₂
Melting Pt: 295…300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6