Antimony(V) chloride 99.995%
Supplier: Apollo Scientific
|
Danger
|
IN1114-250G
IN1114-50G
APOSIN1114-250GEA
1870
EUR
APOSIN1114-250G
APOSIN1114-50G
Antimony(V) chloride 99.995%
Antimony (V) chloride
Formula:
Cl₅Sb Boiling Pt: 79 °C/22mmHg Melting Pt: 2.8 °C Density: 2.358 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011213 CAS Number: 7647-18-9 EINECS: 231-601-8 UN: 1730 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...