Antimony(III) oxide 99,9%
Supplier: Apollo Scientific
Warning
IN1126-250G IN1126-500G IN1126-1000G
APOSIN1126-250GEA 574 EUR
APOSIN1126-250G APOSIN1126-500G APOSIN1126-1000G
Antimony(III) oxide 99,9%
Antimony (III) oxide
Formula: O₃Sb₂
Boiling Pt: 1550 °C (subl.)
Melting Pt: 656 °C
Density: 5.67 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011214
CAS Number: 1309-64-4
UN: 1549
ADR: 6.1,III