Barium nitrate 99.999%
Supplier: Apollo Scientific
Danger
IN1210-100G IN1210-25G
APOSIN1210-100GEA 364 EUR
APOSIN1210-100G APOSIN1210-25G
Barium nitrate 99.999%
Barium nitrate
Formula: Ba(NO₃)₂
MW: 261.34 g/mol
Melting Pt: 592 °C (dec.)
Density: 3.23 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
UN: 1479
ADR: 5.1 I
Merck Index: 13,00986