Bismuth(III) chloride 99.999%
Supplier: Apollo Scientific
Warning
IN1240-100G IN1240-25G IN1240-250G
APOSIN1240-100GEA 683 EUR
APOSIN1240-100G APOSIN1240-25G APOSIN1240-250G
Bismuth(III) chloride 99.999%
Bismuth (III) chloride
Formula: BiCl₃
Boiling Pt: 430 °C
Melting Pt: 230…232 °C
Density: 4.75 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003461
CAS Number: 7787-60-2
EINECS: 232-123-2