Bismuth(III) fluoride, anhydrous 99.99%
Supplier: Apollo Scientific
Danger
IN1243-100G IN1243-25G
APOSIN1243-100GEA 770 EUR
APOSIN1243-100G APOSIN1243-25G
Bismuth(III) fluoride, anhydrous 99.99%
Bismuth (III) fluoride
Formula: BiF₃
MW: 265.98 g/mol
Boiling Pt: 230
Melting Pt: 727 °C
Density: 8.3 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010893
CAS Number: 7787-61-3
UN: 3260
ADR: 8 II
Merck Index: 12,01305