Bismuth(III) oxide 99.999%
Supplier: Apollo Scientific
Warning
IN1255-100G IN1255-500G
APOSIN1255-100GEA 161 EUR
APOSIN1255-100G APOSIN1255-500G
Bismuth(III) oxide 99.999%
Bismuth (III) oxide
Formula: BiO₃
Boiling Pt: 1890 °C
Melting Pt: 817 °C
Density: 8.9 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003462
CAS Number: 1304-76-3