Boron phosphate 99,9%
Supplier: Apollo Scientific
Warning
IN1296-100G IN1296-500G
APOSIN1296-100GEA 319 EUR
APOSIN1296-100G APOSIN1296-500G
Boron phosphate 99,9%
Boron phosphate
Formula: BO₄P
Melting Pt: >1400 °C
Density: 2.81 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011318
CAS Number: 13308-51-5