Cesium carbonate 99%
Supplier: Apollo Scientific
Warning
IN1435-5G IN1435-50G
APOSIN1435-5GEA 75.8 EUR
APOSIN1435-5G APOSIN1435-50G
Cesium carbonate 99%
Cesium carbonate
Formula: Cs₂CO₃
Melting Pt: 610 °C (dec.)
Density: 4.072 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9