Copper(I) chloride 99%
Supplier: Apollo Scientific
Warning
Synonyms: Cuprous chloride, Copper chloride
IN1542-100G IN1542-500G IN1542-1KG
APOSIN1542-100GEA 46.4 EUR
APOSIN1542-100G APOSIN1542-500G APOSIN1542-1KG
Copper(I) chloride 99%
Copper (I) chloride
Formula: ClCu
MW: 99 g/mol
Boiling Pt: 1490 °C
Melting Pt: 430 °C
Density: 4.14 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8 III
Merck Index: 13,02690