Copper(II) chloride dihydrate ACS
Supplier: Apollo Scientific
Danger
APOSIN1543-2.5KGEA 386 EUR
APOSIN1543-2.5KG APOSIN1543-500G APOSIN1543-100G
Copper(II) chloride dihydrate ACS
Copper(II) chloride dihydrate
Formula: CuCl₂·2H₂O
MW: 170,48 g/mol
Melting Pt: 100 °C (dec.)
Density: 2,54 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III