Copper(II) chloride, anhydrous 99,9%
Supplier: Apollo Scientific
Danger
IN1546-1KG IN1546-250G
APOSIN1546-1KGEA 538 EUR
APOSIN1546-1KG APOSIN1546-250G
Copper(II) chloride, anhydrous 99,9%
Copper (II) chloride
Formula: Cl₂Cu
Melting Pt: 620 °C
Density: 3.386 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010972
CAS Number: 7447-39-4
EINECS: 231-210-2
UN: 2802
ADR: 8,III