Dysprosium 99,9%, chips
Supplier: Apollo Scientific
Danger
IN1612-10G IN1612-50G
APOSIN1612-10GEA 139 EUR
APOSIN1612-10G APOSIN1612-50G
Dysprosium 99,9%, chips
Dysprosium
Formula: Dy
Boiling Pt: 2335 °C
Melting Pt: 1409 °C
Density: 8.54 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
UN: 3089
ADR: 4.1,II