Iodine(V) oxide 99%
Supplier: Apollo Scientific
Danger
IN2029-100G IN2029-25G
APOSIN2029-100GEA 203 EUR
APOSIN2029-100G APOSIN2029-25G
Iodine(V) oxide 99%
Iodine (V) oxide
Formula: I₂O₅
Melting Pt: 275…350 °C (dec.)
Density: 5.08 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011356
CAS Number: 12029-98-0
UN: 3085
ADR: 5.1+8,II