Lithium hydroxide monohydrate 98%
Supplier: Apollo Scientific
Danger
IN2340-2.5KG IN2340-500G
APOSIN2340-2.5KGEA 1210 EUR
APOSIN2340-2.5KG APOSIN2340-500G
Lithium hydroxide monohydrate 98%
Lithium hydroxide monohydrate
Formula: LiOH·1H₂O
Boiling Pt: 924 °C
Melting Pt: 462 °C
Density: 1.51 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8,II