Neodymium 99,9%, powder –40 mesh
Supplier: Apollo Scientific
Danger
IN2620-10G IN2620-50G
APOSIN2620-10GEA 625 EUR
APOSIN2620-10G APOSIN2620-50G
Neodymium 99,9%, powder –40 mesh
Neodymium
Formula: Nd
MW: 144.24 g/mol
Boiling Pt: 3074 °C
Melting Pt: 1021 °C
Density: 7 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011130
CAS Number: 7440-00-8
EINECS: 231-109-3
UN: 3089
ADR: 4.1 II