Nickel(II) bromide, anhydrous 99%
Supplier: Apollo Scientific
Danger
Synonyms: Nickel dibromide
IN2674-250G IN2674-50G
APOSIN2674-250GEA 603 EUR
APOSIN2674-250G APOSIN2674-50G
Nickel(II) bromide, anhydrous 99%
Nickel (II) bromide
Formula: Br₂Ni
MW: 218.5 g/mol
Melting Pt: 963 °C
Density: 5.098 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011141
CAS Number: 13462-88-9
UN: 3077
ADR: 9+EHS III
Merck Index: 12,06585