Nickel(II) nitrate hexahydrate 99.995%
Supplier: Apollo Scientific
Danger
IN2698-250G IN2698-50G
APOSIN2698-250GEA 290 EUR
APOSIN2698-250G APOSIN2698-50G
Nickel(II) nitrate hexahydrate 99.995%
Nickel (II) nitrate hexahydrate
Formula: Ni(NO₃)₂·6H₂O
MW: 290.79 g/mol
Boiling Pt: 137 °C
Melting Pt: 56.7 °C
Density: 2.05 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149805
CAS Number: 13478-00-7
EINECS: 236-068-5
UN: 2725
ADR: 5.1 III
Merck Index: 12,06596