Niobium 99,8%, powder –60 mesh
Supplier: Apollo Scientific
IN2731-100G IN2731-25G
APOSIN2731-100GEA 101 EUR
APOSIN2731-100G APOSIN2731-25G
Niobium 99,8%, powder –60 mesh
Niobium
Formula: Nb
Boiling Pt: 4742 °C
Melting Pt: 2468 °C
Density: 8.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1