Periodic acid 99+%
Supplier: Apollo Scientific
Danger
Synonyms: ortho-Periodic acid, Orthoperiodic acid
IN2845-5G IN2845-50G
APOSIN2845-5GEA 43.5 EUR
APOSIN2845-5G APOSIN2845-50G
Periodic acid 99+%
Periodic acid
Formula: H₅IO₆
Boiling Pt: 140 °C
Melting Pt: 122…130 °C
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011344
CAS Number: 10450-60-9
EINECS: 233-937-0
UN: 3085
ADR: 5.1+8,II