Tantalum 99,98%, powder –60 mesh
Supplier: Apollo Scientific
IN3400-100G IN3400-25G
APOSIN3400-100GEA 0 EUR
APOSIN3400-100G APOSIN3400-25G
Tantalum 99,98%, powder –60 mesh
Tantalum
Formula: Ta
Boiling Pt: 5429 °C
Melting Pt: 2996 °C
Density: 16.6 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7