Tin(II) chloride, anhydrous 98%
Supplier: Apollo Scientific
Danger
Synonyms: Stannous chloride, Tin dichloride
IN3644-100G IN3644-2.5KG IN3644-500G
APOSIN3644-100GEA 79.9 EUR
APOSIN3644-100G APOSIN3644-2.5KG APOSIN3644-500G
Tin(II) chloride, anhydrous 98%
Tin (II) chloride
Formula: SnCl₂
Boiling Pt: 652 °C
Melting Pt: 247 °C
Density: 3.95 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011241
CAS Number: 7772-99-8
EINECS: 231-868-0
UN: 3260
ADR: 8,III