Zinc oxide 99,9%
Supplier: Apollo Scientific
Warning
Synonyms: Zinci oxidum, Zinc (II) oxide
IN3888-1KG IN3888-250G IN3888-5KG
APOSIN3888-1KGEA 151 EUR
APOSIN3888-1KG APOSIN3888-250G APOSIN3888-5KG
Zinc oxide 99,9%
Zinc oxide
Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.606 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
UN: 3077
ADR: 9+EHS III