Zirconium 99,8%, lumps
Supplier: Apollo Scientific
Danger
IN3898-100G IN3898-25G
APOSIN3898-100GEA 0 EUR
APOSIN3898-100G APOSIN3898-25G
Zirconium 99,8%, lumps
Zirconium
Formula: Zr
MW: 91.22 g/mol
Boiling Pt: 3580 °C
Melting Pt: 1852 °C
Density: 6.506 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9