Zirconium tetrachloride 99,5%
Supplier: Apollo Scientific
Danger
Synonyms: Zirconium (IV) chloride
IN3907-100G IN3907-500G
APOSIN3907-100GEA 169 EUR
APOSIN3907-100G APOSIN3907-500G
Zirconium tetrachloride 99,5%
Zirconium tetrachloride
Formula: Cl₄Zr
MW: 233.03 g/mol
Boiling Pt: 331 °C
Melting Pt: 300 °C (subl.)
Density: 2.81 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011306
CAS Number: 10026-11-6
UN: 2503
ADR: 8 III
Merck Index: 12,10307